Order Entry
Mexico
Orders LinkContactUsLinkComponent
BBL™ Mannitol Salt Agar, BD

BBL™ Mannitol Salt Agar, BD

Mannitol Salt Agar, 500 g
Catalog # 90000-182
Supplier #:  211407
BBL™ Mannitol Salt Agar, BD
Catalog # 90000-182
CAS Number:  
BBL™ Mannitol Salt Agar, BD
BBL™ Mannitol Salt Agar, BD

Mannitol Salt Agar, 500 g

CAS Number:  
Price
$2,670.00
Log in for availability
Each (500g)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Mannitol Salt Agar is a selective and differential medium for the detection and enumeration of staphylococci from clinical and nonclinical specimens.

  • Beef extract and peptones provide essential growth factors
  • 7.5% concentration of sodium chloride inhibits bacterial growth
  • Mannitol fermentation aids in differentiation of staphylococcal species

Difference in colony growth colors also acts as an identifying feature.

Documents

Specifications

Specifications

  • Description:
    Mannitol Salt Agar
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.