Beaker Tri pour
Catalog # 470142-552
Supplier: DYN-A-MED PRODUCTS
Specifications
- Capacity:800 ml (27 oz.)
- Description:Beaker Tri Pour 800 mL, Pkg./10
- Cat. no.:470142-552
- Supplier No.:80097
Specifications
About this item
This tri-pour beaker is perfect for any classroom. The 800ml beakers are extremely durable and portable. They make science easy and the polypropylene material is safe to use.