Micro Titertubes, Thermo Scientific
1.2 ml, PP, Natural, Micro Titertube, Robotic Rack
Catalog # 46300-506
Supplier: Thermo Fisher Scientific
Supplier #: RC-845-TP
Jump to:
Product Details & Documents
Micro Titertubes offer additional volume capacity for sample dilution and/or mixing prior to transfer into micro titer plates.
- Pre-racked in an 8×12 format
- Available plain or with frosted writing areas
- Autoclaved
- Sealed and frozen for storage
Ideal for a wide variety of laboratory procedures such as HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, blood bank sample freezer storage or transport of specimens and reagents.
Specifications
- Capacity:1.2 ml
- Color:Natural
- Description:Micro Titertube
- Material:PP
- Packaging:Robotic Rack