Order Entry
Mexico
ContactUsLinkComponent
 
Color Standard Solution, Hach
Catalog # 10000-830
Proveedor:  Avantor
Número CAS:  
Color Standard Solution, Hach
Catalog # 10000-830
Proveedor:  Avantor
Número CAS:  

Especificaciones

  • Concentration:
    500 platinum cobalt units
  • Description:
    Standard Color Solution, 1 L bottle
  • Size:
    1 L
  • Cat. no.:
    10000-830
  • Supplier No.:
    141453

Especificaciones

Acerca de este artículo

For accuracy checks in Color determinations.

  • Simple accuracy checks
  • Minimize waste
  • Reliable results
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.