PYREX® 75° Angled Tube Adapter with Two-Way 24/40 Standard Taper Joints, Corning
89091-524
: Corning
Product available on GSA Advantage®
- Description:PYREX® 75° Angled Tube Adapter with Two-Way [ST] Joints
- Environmentally Preferable:Low Manufacturing Impact
- Cat. no.:89091-524
- ST Joint Size:24/40
- Supplier No.:8920-24
- Environmental certification:I-RECs, ISO 14001 - Monterrey
These angled two-way adapters are made of PYREX® borosilicate glass.
- Arms are approximately 75° apart
- Both ends are equipped with an inner 24/40 Standard Taper joint
It is often used as a stillhead adapter.