Order Entry
Mexico
ContactUsLinkComponent
Econo™ Sterile Knowles Bandage Scissors, Floor Grade, Sklar
Econo™ Sterile Knowles Bandage Scissors, Floor Grade, Sklar
Catalog # 89023-746
Supplier:  SKLAR CORPORATION
undefined
Econo™ Sterile Knowles Bandage Scissors, Floor Grade, Sklar
Catalog # 89023-746
Supplier:  SKLAR CORPORATION
Supplier Number:  96-2402
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Econo™ Sterile Knowles Bandage Scissors
  • Length:
    13.9 cm (5½")
  • Material:
    Stainless steel
  • Curvature:
    Angled
  • Cat. no.:
    89023-746
  • Supplier No.:
    96-2402
  • Blade Type:
    Blunt/Blunt
Specifications
About this item
Econo™ Sterile Knowles Bandage Scissors are durable, lightweight, and designed for cutting thick dressing material
Bandage scissors have a blunt tip on the upper blade so as not to cause any injury to the patient while removing bandages or dressings. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination. They are available in bulk quantities. Scissors are angled with smooth, blunt/blunt tips.