Order Entry
Mexico
Orders LinkContactUsLinkComponent
Friction Box
Friction Box
Friction Box
Catalog # 470219-712
Supplier #:  FRBX01
Friction Box
Catalog # 470219-712
Friction Box
Friction Box
Friction Box
Clearance
$5.60
Sale
Valid until 31/12/2026
Log in for availability
Each (1UN)
Product Details & Documents
Sturdy box for investigating frictional forces.
  • Smooth wood
  • Holds up to 2 kg
This friction box is used for coefficient of friction studies when larger loads are required. The 6" x 3.5" x 2.5" box will accommodate a load of 2kg or more. Smooth sanded wood with two leather strips attached to the bottom.
Shipping weight: 0.5 lbs. Dimensions: 7.5" x 4" x 3".
Specifications
  • Description:
    Friction Box