Knife-Edge Clamp
Knife-Edge Clamp
Catalog # 470219-710
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: CLMKE1
Jump to:
Product Details & Documents
This clamp can be attached to any standard meter stick. It is used as a knife-edge fulcrum for supporting the meter stick in the inverted position.
- Fits standard meter sticks
- Thumbscrew to hold securely
The knife edge clamp is useful when invetsigating torque on a lever.
Specifications
- Description:Knife-Edge Clamp