Order Entry
Mexico
ContactUsLinkComponent
Conductors; Set of Three
Conductors; Set of Three
Catalog # 470219-522
undefined
Conductors; Set of Three
Catalog # 470219-522
Supplier Number:  PH0880
Specifications
  • Description:
    Conductors; Set of Three
  • Cat. no.:
    470219-522
  • Supplier No.:
    PH0880
Specifications
About this item
Made of brass, interchangeable on pillars, fitted with insulators of ebonite on base. Spheres 5cm diameter, Cylinder 12.5 x 5cm diameter, Cone 10 x 5cm length x diameter.