Urea 99.0-100.5% ACS
Catalog # BDH4602-500G
Supplier: VWR
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle
- Size:500 g
- Cat. no.:BDH4602-500G
- Supplier No.:BDHVBDH4602-500G
Specifications
Specification Test Results
- Assay:99.0 - 100.5 %
- Melting point (132-135°C):Passes test
- Heavy metals (as Pb):Max. 0.001 %
- Insoluble matter:Max. 0.01 %
- Residue after ignition:Max. 0.01 %
- Cl (Chloride):Max. 5 ppm
- SO4 (Sulphate):Max. 0.001 %
- Fe (Iron):Max. 0.001 %
Specification Test Results
About this item
:
Merck Index:13,09935
Boiling Pt:197 °C (1013 hPa)
Boiling Pt:197 °C (1013 hPa)
Melting Pt:132.7 °C
Density:1.34 g/cm³ (20 °C)
:60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
REACH:01-2119463277-33
CAS Number:57-13-6
Formula:NH₂CONH₂
MDL Number:MFCD00008022