Order Entry
Mexico
ContactUsLinkComponent
TouchNTuff® 92-605 Nitrile gloves, Ansell
  10769-516
undefined
TouchNTuff® 92-605 Nitrile gloves, Ansell
  10769-516
 :  588081
  • Glove Color:
    Green
  • Glove size:
    M
  • Ambidextrous / hand-specific:
    Ambidextrous
  • Length:
    30 cm (11¹³⁄₁₆")
  • Environmentally Preferable:
  • Finger thickness:
    7.9 mil
  • Palm thickness:
    4.7 mil
  • Cuff thickness:
    3.1 mil
  • Cat. no.:
    10769-516
  • Supplier No.:
    114097
The TouchNTuff 92-605 gloves offer proven splash resistance against hazardous chemicals.
  • Non sterile
  • Ambidextrous
  • Strong grip in wet or dry conditions
It resists a greater variety of industrial chemicals for longer periods than any other nitrile disposable gloves.
For use in the agriculture and viticulture, automotive and transportation, building and construction and chemical industries for all final stages of meat or fish processing, controlled-environment rooms, prepared meals, dairy processing and catering.
Manufactured from nitrile, TouchNTuff offers up to four times the puncture resistance of comparable natural rubber latex, and three times the resistance of similar neoprene gloves. It contains no natural rubber latex proteins, which prevents Type I allergies for the wearer. Primary skin irritation studies and Insult Patch tests have also shown no evidence of risk from irritation or allergic contact dermatitis. With a unique 'Thin Nitrile Technology' formulation, this glove offers easy donning and strong grip in wet or dry conditions. The glove is versatile and suitable for many different uses. Inspected to an AQL of 1.5 for pinholes, which matches the standards for medical gloves.
 : EN 420, EN374/1, 2, 3, ASTM D 6319, FDA 21 CFR 177-2600, Areete 09/11/04, European Regulation 1935/2004, German Recommendation for Food Commodities.