Examination Gloves, Microflex®, Ansell
About this item
This thicker version of the popular Microflex® Cobalt® X N21 glove offers fit and feel with the extra protection of a thicker glove.
- Fully textured to provide grip in any condition
- Non sterile
- Ambidextrous
- Gloves are blue and feature beaded cuffs
For use in life science, electronics, and food processing industries for analytical testing/measurements; assembly and inspection; biotechnologies; cleaning; food processing; handling small parts; laboratory analysis; laboratory testing; light duty maintenance and clean up; light assembly tasks; maintaining equipment and instruments; pharmaceutical manufacturing; preparing pharmaceutical products; sanitation and dish washing; and as an under glove for meat/fish processing.
3 Options Available Below
Product Details & Documents
This thicker version of the popular Microflex® Cobalt® X N21 glove offers fit and feel with the extra protection of a thicker glove.
- Fully textured to provide grip in any condition
- Non sterile
- Ambidextrous
- Gloves are blue and feature beaded cuffs
For use in life science, electronics, and food processing industries for analytical testing/measurements; assembly and inspection; biotechnologies; cleaning; food processing; handling small parts; laboratory analysis; laboratory testing; light duty maintenance and clean up; light assembly tasks; maintaining equipment and instruments; pharmaceutical manufacturing; preparing pharmaceutical products; sanitation and dish washing; and as an under glove for meat/fish processing.
Specifications for Product Family
Specifications
- Catalog No:
- 10770-436
- 10770-460
- 10770-462
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Cuff style:
- Beaded
- Beaded
- Beaded
- Cuff thickness:
- 3.9 mil
- 3.9 mil
- 3.9 mil
- Finger thickness:
- 5.5 mil
- 5.5 mil
- 5.5 mil
- Palm thickness:
- 4.7 mil
- 4.7 mil
- 4.7 mil
Specifications
Product Family Options
Product Information
- Glove sizeGlove ColorLengthAvailabilityPrice
- SBlue24 cm (9⅜")Paquete de 100$347.00Case de 10 (100UN)$419.90Supplier Number: N211Specifications:
More
Specifications
Cat. No.10770-436Ambidextrous / hand-specificAmbidextrousCuff styleBeadedCuff thickness3.9 milFinger thickness5.5 milPalm thickness4.7 mil - MBlue24 cm (9⅜")Paquete de 100$347.00Case de 10 (100UN)$419.90Supplier Number: N212Specifications:
More
Specifications
Cat. No.10770-460Ambidextrous / hand-specificAmbidextrousCuff styleBeadedCuff thickness3.9 milFinger thickness5.5 milPalm thickness4.7 mil - LBlue24 cm (9⅜")Paquete de 100$347.00Case de 10 (100UN)$419.90Supplier Number: N213Specifications:
More
Specifications
Cat. No.10770-462Ambidextrous / hand-specificAmbidextrousCuff styleBeadedCuff thickness3.9 milFinger thickness5.5 milPalm thickness4.7 mil