Accessories for Overhead Stirrers, Caframo
77776-350
- Description:Fastening Clamp for Overhead Stirrers
- Cat. no.:77776-350
- Supplier No.:A120
The Caframo Fastening Clamp attaches the overhead stirrer to the stand
It is composed of a cast zinc aluminum alloy with chemical resistant epoxy coating. The clamp accepts a 1/2" diameter stirrer support rod and stand rods between 0.6 and 1.2" in diameter. Handles are composed of 304 stainless steel.