Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Analytical Balances
Ward's® Analytical Balances
Supplier:   OHAUS CORP.
Ward's® Analytical Balances
Supplier:  OHAUS CORP.
Certificates
Ward's® Analytical Balances
Ward's® Analytical Balances
3 Options Available Below
Product Details & Documents
These balances are perfect for school laboratory, AP class, or prep-room use.
  • Easy-to-read LCD display
  • One-year manufacturer's warranty
  • ABS top housing
  • Removable stainless steel pan
  • Integrated weigh below hook
Designed for use in an instructional environment, these balances deliver dependable mass measurements for solution preparation, quantitative analysis, and more.
All models come with a weighing chamber draft shield. Weighing modes include grams, milligrams, kilograms and Newtons. An RS-232C interface is available (cord sold separately).
Simple operation and durable construction make them ideal for student use.
Ordering information: One-year manufacturer's warranty is included with purchase.
Product Family Options
Product Information
  • Weighing Capacity
    Linearity
    Pan/Platform Size
    Readability
    Repeatabilty
    Availability
    Price
  • 62 g
    0.0002 g
    Ø 90 mm
    0.0001 g
    0.0001 g
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $65,058.87
    $65,058.87
    Each (1UN)
       $65,058.87
    Supplier Number: 30499238
  • 120 g
    0.0002 g
    Ø 90 mm
    0.0001 g
    0.0001 g
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $66,428.72
    $66,428.72
    Each (1UN)
       $66,428.72
    Supplier Number: 30499239
  • 220 g
    0.0002 g
    Ø 90 mm
    0.0001 g
    0.0001 g
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $78,559.94
    $78,559.94
    Each (1UN)
       $78,559.94
    Supplier Number: 30499240