Order Entry
Mexico
ContactUsLinkComponent
Sterile Flocked Swab, with Molded Plastic Applicator & Nylon Fiber Tip
  10755-188
undefined
Sterile Flocked Swab, with Molded Plastic Applicator & Nylon Fiber Tip
  10755-188
 :  520CS01.BX
  • Description:
    Flocked Swab, with 30 mm Breakpoint from Swab Tip
  • Cat. no.:
    10755-188
  • Packaging:
    Peel Pouch
  • Supplier No.:
    520CS01.BX
Flocked swabs absorb and immediately release a sample into liquid media
COPAN swabs are compatible with multiple applications and platforms, such as rapid antigen testing, EIA, molecular-based assays, DFA, cytology testing, forensics, bacteriology, and virology culture.