Order Entry
Mexico
ContactUsLinkComponent
 
Whatman™ Cellulose Nitrate Membranes
 :  Cytiva
 
undefined
Whatman™ Cellulose Nitrate Membranes
 :  Cytiva
 

 

  • Catalog No:
  • 28297-778
  • 28454-042
  • 10034-896
  • 28159-547
  • 28152-825
  • 10147-926
  • 28159-570
  • 28454-079
  • 28454-053
  • 28454-097
  • 28454-089
  • 10035-534
  • Type:
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Hydrophilic
  • Weight:
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • 3.6 mg/cm² Min. to 5.5 mg/cm² Max.
  • Max. operating temperature:
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • Application:
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Sample preparation
    Microbiological studies
    Filtration of aqueous solutions
  • Autoclavable:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Colour:
  • White
  • White
  • White
  • White
  • White
  • White
  • White
  • White
  • White
  • White
  • White
  • White
  • Porosity:
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.
  • 66% Min. to 84% Max.

 

Frequently Bought Together