

2-Propanol ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
Catalog # BJLC323-2.5
Supplier: HONEYWELL INTERNATIONAL INC MX
CAS Number:
Specifications
- Pack type:Glass bottle, GL45
- Size:2.5 L
- Cat. no.:BJLC323-2.5
- Supplier No.:LC323-2.5
Specifications
Specification Test Results
- GC Analysis:99.9 % min.
- LC-MS Negative Mode (as 4-nitrophenol):50 ppb max.
- LC-MS Positive Mode (as reserpine):50 ppb max.
- Refractive Index (20ºC):1.3766 - 1.3778
- Residue:1 mg/L max.
- Trace Metals:Passes test
- UV Cutoff:205 nm max.
- Water by Karl Fischer Titration:0.05 % max.
Specification Test Results
About this item
:
:60.1 g/mol
Boiling Pt:12.0 %(V)
Melting Pt:–88 °C
Density:0.785 g/cm³ (20 °C)
Flash Pt:12 °C
UN:1219
ADR:3,II
:GHS07
:GHS02
Storage Temperature:Ambient
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674