SP Wilmad-LabGlass Tube for Stalagmometer, SP Industries
Catálogo # 14203-776
Proveedor: SP Industries
Especificaciones
- Capacity:2.5 ml
- Description:Stalagmometer Tube
- Environmentally Preferable:
- Cat. no.:14203-776
- Supplier No.:LG-5050-102
Especificaciones
Acerca de este artículo
Specifically designed for checking the surface tension of liquids, each tube is accurately calibrated to quickly determine surface tension by the drop count method.
- Graduated above and below the bulb
This tube allow for flexibility in sample size of most liquid baths.
Entrega: All tubes are supplied with instructions for use.