Order Entry
Mexico
Orders LinkContactUsLinkComponent
Precision Specific Gravity Hydrometers, Kimble Chase
Precision Specific Gravity Hydrometers, Kimble Chase
Precision Specific Gravity Hydrometers, Kimble Chase
Precision Specific Gravity Hydrometers, Kimble Chase
Serial number is printed on the scale and a certificate of traceability is included
Model 52110-2000 is a set comprising one each of all 8 precision specific gravity hydrometers.
The hydrometer meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy. The certificate of traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards. The ballast is made from steel pellets and a binder, and it is free of heavier metals.
Product Family Options
  • Description
    Sample Volume
    Length
    Body O.D.
    Stem O.D.
    Graduation Interval
    Graduation Range
  • Hydrometer
    300 ml
    310 mm
    16 mm
    7 mm
    0.001 SG
    SG 0.700–0.810
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
     : 52110-0708
  • Hydrometer
    500 ml
    300 mm
    18 mm
    7 mm
    0.001 SG
    SG 0.800–0.910
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
     : 52110-0809
  • Hydrometer
    500 ml
    310 mm
    18 mm
    7 mm
    0.001 SG
    SG 0.900–1.010
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
     : 52110-0910
  • Hydrometer
    500 ml
    300 mm
    16 mm
    8 mm
    0.002 SG
    SG 1.000–1.220
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
     : 52110-1012
  • Hydrometer
    300 ml
    300 mm
    16 mm
    7 mm
    0.002 SG
    SG 1.200–1.420
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
     : 52110-1214
  • Hydrometer
    500 ml
    300 mm
    18 mm
    7 mm
    0.002 SG
    SG 1.400–1.620
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
     : 52110-1416
  • Hydrometer
    500 ml
    300 mm
    18 mm
    7 mm
    0.002 SG
    SG 1.600–1.820
    Each (1UN)
    Each (1UN) - $1,258.63
    $1,258.63
    Each (1UN)
       $1,258.63
     : 52110-1618
  • Hydrometer Set
    Each (1UN)
    Each (1UN) - $18,691.06
    $18,691.06
    Each (1UN)
       $18,691.06
     : 52110-2000