Cannon-Fenske Routine Viscometer, Cannon Instrument Company
Supplier: CANNON INSTRUMENT CO
Certificates
About this item
The Cannon-Fenske Routine Viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104
Available calibrated (CFRC) and uncalibrated (CFRU). Minimum sample volume 7 mL. Minimum bath depth 8 inches.
Frequently Bought Together