Order Entry
Mexico
Orders LinkContactUsLinkComponent
Kerr Effect
Kerr Effect
Kerr Effect with Halogen lamp
Catalog # 470163-958
Supplier #:  SK019
Kerr Effect
Catalog # 470163-958
Kerr Effect
Kerr Effect
Kerr Effect with Halogen lamp
Clearance
$1,202.20
Sale
Valid until 31/12/2026
Log in for availability
Each (1UN)
Product Details & Documents
Understand the Kerr effect like you never have before!
  • Demonstrate the Kerr effect in nitrobenzene solution
  • Measure the light intensity using photo detector
This apparatus is used to demonstrate the Kerr effect in nitrobenzene solution and to measure the light intensity as a function of voltage across the Kerr cell using photo detector.
Salient features:
All connections are made using safety sockets.
Principle and Working:
In the present setup, a small glass container mounted with plate capacitor is filled with liquid. The container is kept between two filters placed at 90°. There will be dark field of view on screen initially but after applying electric field, the field of view brightens, because the light beam becomes elliptically polarized when passed through the birefringent liquid.
Included:
Kerr Cell
Polarizer / Analyzer 2
Object screen
Prism table
Optical Bench
Flexible plug lead,1m (red)
Flexible plug lead,1m (black)
Transverse Saddle
Halogen Light Source (for halogen version)
Convex lens in holder(for halogen version)
Monochromatic light filter(for halogen version)
Diode Laser(for laser version)
Photodetector(for laser version)
Specifications
  • Description:
    Kerr Effect with Halogen lamp