Order Entry
Mexico
Orders LinkContactUsLinkComponent
Flame Test Station Kit
Flame Test Station Kit
Flame Test Lab Station Kit, Instructions, 100
  470013-550
 :   Avantor
Flame Test Station Kit
  470013-550
 :  Avantor
Flame Test Station Kit
Flame Test Station Kit
Flame Test Lab Station Kit, Instructions, 100
$1,339.80
Each (1UN)
Setting Up Your Flame Test Lab Just Got Easier.
  • Kit contains all necessary materials for lab set up
  • Further experiments can be performed with spectroscopes, available separately
  • Solutions include: calcium nitrate, copper(II) nitrate, potassium nitrate, sodium nitrate, and strontium nitrate
This kit has everything you need, conveniently arranged and clearly explained. Simply open up the five included bottles of solutions, Lay out the wood splints, pass out the instructions and you are on your way to a successful lab activity. Cobalt glass is also included to help distinguish between the potassium and the sodium flame test.
  • Description:
    Flame Test Lab Station Kit
  • Includes:
    Instructions
  • No. of Uses:
    100