- Description:Ink Chromatography and Forensics STEM Kit
- No. of Students:40
- Cat. no.:470213-358
- Supplier No.:AISCRKIT
Students solve the case with ink chromatography and STEM.
- Hands-On STEM Activity
- Model and Inquiry Activities
- Teacher and Student Guides Included
- Material Accommodates up to 40 Students
- Covered Concepts Include Data Analysis, Chromatographs, and More
This kit introduces students to ink chromatography and its application in solving forensics cases. They gain experience in creating ink databases, identifying ‘ink tags’ to ‘date’ various inks and then use them, as professional document examiners do, in solving two mysteries involving document inks—an apparent suicide note and a forgery case.
Covered concepts include data analysis, chromatographs, inks and their composition-mixtures, compounds, mixtures, molecules, solubility, affinity, polar substances, Rf values, capillary action, light energy, fluorescence, and hypothesis testing.