Order Entry
Mexico
Orders LinkContactUsLinkComponent
Miniature LED Lights
Minature LED bulb, pack of 10, E10 base, 3.2 V
  470224-034
 :  OPDT-W4DK86A1B
Miniature LED Lights
  470224-034
Miniature LED Lights
Miniature LED Lights
Minature LED bulb, pack of 10, E10 base, 3.2 V
$195.00
Paquete de 10
These lights are great to use with a variety of electricity and science projects.
  • Your batteries will last longer: eco-friendly LEDs draw only 25 mA
  • Bulbs light only when current is in correct direction
  • Size E10 screw base
  • Pack of 10
Perfect for student investigations of DC circuits. Not recommended to exceed 3 V. The bulbs light only when the current is in the correct direction.
 : One package has 10 bulbs.
  • Description:
    Minature LED bulb, pack of 10, E10 base, 3.2 V