Order Entry
Mexico
Orders LinkContactUsLinkComponent
Singing Rods
Singing Rods
Singing Rods
Catalog # 470163-896
Supplier #:  3637-00
Singing Rods
Catalog # 470163-896
Singing Rods
Singing Rods
Singing Rods
Price
$475.98
Log in for availability
Each (1UN)
Product Details & Documents
A must for explorations of sound and waves.
  • Sold as a set of two
  • Approx length: 20" (500mm) and 30" (750mm)
  • Comes with a Teacher's Guide with detailed instructions and a bag of crushed rosin to lightly coat your fingertips
These rods are an easy way to demonstrate longitudinal waves as opposed to transverse waves. They're ideal for teaching about nodes and anti-nodes in standing waves.
Specifications
  • Description:
    Singing Rods