Set of 5 Single Slits
Catalog # 470163-770
Supplier: AMERICAN 3B SCIENTIFIC SE
Specifications
- Description:Set of 5 Single Slits
- Cat. no.:470163-770
- Supplier No.:U8470790
Specifications
About this item
Five single slits of different widths in a slide frame.
- Slit widths: 0.1/ 0.2/ 0.4/ 0.8/ 1.6 mm
- Dimensions: 50x50 mm²