High Speed Gearbox
High Speed gearbox(11.6:1 & 18:1)
Catalog # 470096-268
Supplier: TAMIYA AMERICA INC.
Supplier #: 72002
Jump to:
Product Details & Documents
The High Speed Gear Box, High Efficiency, is perfect for high speed applications
Two gear ratios are possible, and several styles of crank arms are supplied for a variety of uses. A RE-260 motor is included. The gear box can be mounted vertically or horizontally.
Specifications
- Description:High Speed gearbox(11.6:1 & 18:1)