Order Entry
Mexico
Orders LinkContactUsLinkComponent
Quantofix Silver Test Strips, CTL Scientific Supply
Silver test strips, 0 - 1 - 2 - 3 - 5 - 7 - 10 g/L
  10217-556
 :  91350
Quantofix Silver Test Strips, CTL Scientific Supply
  10217-556
 :  
Quantofix Silver Test Strips, CTL Scientific Supply
Quantofix Silver Test Strips, CTL Scientific Supply
Silver test strips, 0 - 1 - 2 - 3 - 5 - 7 - 10 g/L
 :  
$1,249.90
Each (1UN)
Quantofix Silver test strips for the semi-quantitative determinaton of silver.
Quantofix Silver has a gradation of scale: 0-1-2-3-5-7-10.
 : Packaged as a box of 100.
  • Description:
    Silver test strips
  • Range:
    0 - 1 - 2 - 3 - 5 - 7 - 10 g/L
  • Cat. No.:
    10217-556
  • Storage:
    RT