Order Entry
Mexico
ContactUsLinkComponent
Flask Stand, PP, United Scientific Supplies
Flask Stand, PP, United Scientific Supplies
Catalog # 470164-044
undefined
Flask Stand, PP, United Scientific Supplies
Catalog # 470164-044
Supplier Number:  45105
Specifications
  • Description:
    Flask Stand, PP
  • Cat. no.:
    470164-044
  • Supplier No.:
    45105
Specifications
About this item
For use with flasks up to 10 L, this polypropylene flask stand is ribbed to safely hold flasks in place.
  • Autoclavable
The flask stand has 12 different steps to hold a variety of flasks centerd and stable.