Order Entry
Mexico
Orders LinkContactUsLinkComponent
Spectroquant® Nitrite Test, MilliporeSigma

Spectroquant® Nitrite Test, MilliporeSigma

Griess' reaction, Prove, Nova 60, Multy, Move, Fiber Case, 335
Catalog # EM1.14776.0002
Supplier:   Avantor
Spectroquant® Nitrite Test, MilliporeSigma
Catalog # EM1.14776.0002
Supplier:  Avantor
CAS Number:  
Spectroquant® Nitrite Test, MilliporeSigma

Griess' reaction, Prove, Nova 60, Multy, Move, Fiber Case, 335

CAS Number:  
Price
$6,977.79
Log in for availability
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Documents

Specifications

Specifications

  • Chemical method:
    Griess' reaction
  • Pack type:
    Fiber Case
  • No. of Tests:
    335
  • For Unit:
    Prove, Nova 60, Multy, Move
  • Cat. No.:
    EM1.14776.0002
  • Storage temperature:
    Store at +15°C to +25°C.
  • Method:
    Photometric

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.