Fluoride Cell Test, Spectroquant®, MilliporeSigma
Alizarin complexone, Prove, Nova 60, Multy, Move, 0.10 - 1.80 mg/L F⁻, 25
Supplier: MERCK, S.A. DE C.V. SV
Supplier #: 1.00809.0001
Fluoride Cell Test, Spectroquant®, MilliporeSigma
Catalog # EM1.00809.0001
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Chemical method:Alizarin complexone
- Range:0.10 - 1.80 mg/L F⁻
- No. of Tests:25
- For Unit:Prove, Nova 60, Multy, Move
- Cat. No.:EM1.00809.0001
- Storage temperature:Store at +15°C to +25°C.
- Method:Photometric 0.10 - 1.80 mg/l F⁻
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



