Order Entry
Mexico
Orders LinkContactUsLinkComponent
Spectroquant® Nitrite Cell Test, Photometric Method, MilliporeSigma

Spectroquant® Nitrite Cell Test, Photometric Method, MilliporeSigma

1 pc, Iron sulfate, Prove, Nova 30/60, Multy, Move, 90.0 mg/L NO₂-N 3.3 - 295.2 mg/L NO₂⁻, 25
Catalog # EM1.00609.0001
Supplier #:  EM1.00609.0001
Spectroquant® Nitrite Cell Test, Photometric Method, MilliporeSigma
Catalog # EM1.00609.0001
CAS Number:  
Spectroquant® Nitrite Cell Test, Photometric Method, MilliporeSigma

1 pc, Iron sulfate, Prove, Nova 30/60, Multy, Move, 90.0 mg/L NO₂-N 3.3 - 295.2 mg/L NO₂⁻, 25

CAS Number:  
Price
$6,107.88
Log in for availability
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments

  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.

Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).

Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.

Specifications

Specifications

  • Chemical method:
    Iron sulfate
  • Size:
    1 pc
  • Range:
    90.0 mg/L NO₂-N 3.3 - 295.2 mg/L NO₂⁻
  • No. of Tests:
    25
  • For Unit:
    Prove, Nova 30/60, Multy, Move
  • Cat. No.:
    EM1.00609.0001
  • Storage temperature:
    Store at +15°C to +25°C.
  • Method:
    Photometric 1.0 - 90.0 mg/l NO₂-N 3.3 - 295.2 mg/l NO₂⁻

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.