Order Entry
Mexico
ContactUsLinkComponent
MColortest™ Ammonium Test, MilliporeSigma
  EM1.08024.0001
 :  
MColortest™ Ammonium Test, MilliporeSigma
  EM1.08024.0001
 :  1080240001
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Chemical method:
    Indophenol blue
  • Description:
    Ammonium Test
  • Method:
    Colorimetric, indophenol blue, with color card and sliding comparator 0.2 - 0.4 - 0.6 - 1 - 2 - 3 - 5 mg/l NH₄⁺
  • Storage temperature:
    Store at +15°C to +25°C.
  • Cat. no.:
    EM1.08024.0001
  • Range:
    0.2 - 0.4 - 0.6 - 1 - 2 - 3 - 5 mg/L NH₄⁺
  • Supplier No.:
    1.08024.0001
  • No. of Tests:
    50
The test measures both ammonium ions and dissolved ammonia in e.g
groundwater and surface water, seawater, drinking water and aquarium water. The test kit contains the easy to use colour card and the sliding comparator.