To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Especificaciones
- Chemical method:Vanadate-molybdate
- Description:Phosphate Test Refill Pack for 1.18388
- Environmentally Preferable:
- Method:Colorimetric, with color-disk comparator
- Storage temperature:Store at +15°C to +25°C.
- Cat. no.:EM1.18466.0002
- Supplier No.:1.18466.0002
- No. of Tests:300
Especificaciones
Acerca de este artículo
The test can be used for the determination of orthophosphate in groundwater and surface water, drinking water, wastewater
When the reagents of the MColortestTM test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MColortestTM test kits, making it a particularly economical system.
The concentration can be measured semiquantitatively by using the colour comparator.