Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Zinc bromide, anhydrous for synthesis, Sigma-Aldrich®

Glass bottle, 1 kg
  EM8.18631.1000
 
 
 :  8.18631.1000
Zinc bromide, anhydrous for synthesis, Sigma-Aldrich®
  EM8.18631.1000
 :  7699-45-8
Zinc bromide, anhydrous for synthesis, Sigma-Aldrich®

Glass bottle, 1 kg

 :  7699-45-8
 
$11,785.90
   
Each (1kg) Case de 24 (1kg)
 
   
 
 :  Zinc (II) bromide
  •  :7699-45-8
  •  :Br₂Zn
  •  :MFCD00011294
  •  :225.2 g/mol
  •  :8,II
  •  :M-Clarity™ quality level = MQ200
  •  :3260
  •  :8,III

 

 

 

  • Solubility:
    4470 g/l (20 °C)
  • Melting Point:
    394 °C
  • Molar Mass:
    225.2 g/mol
  • Boiling Point:
    697 °C (1013 mbar)
  • Density:
    4.2 g/cm3 (20 °C)
  • pH:
    4 (H2O, 20 °C) (saturated solution)
  • Hill Formula:
    Br2Zn
  • Chemical Formula:
    ZnBr2

 

  • Pack type:
    Glass bottle
  • Size:
    1 kg

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.