Methacrylic acid stabilised for synthesis, Sigma-Aldrich®
Glass Bottle, 500 ml
Supplier: MERCK, S.A. DE C.V. SV
Supplier #: 8005780500
Methacrylic acid stabilised for synthesis, Sigma-Aldrich®
Catalog # EM8.00578.0500
Supplier: MERCK, S.A. DE C.V. SV
CAS Number: 79-41-4
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Synonyms: 2-Methylpropenoic acid
- CAS Number:79-41-4
- Formula:C₄H₆O₂
- MDL Number:MFCD00002651
- Molecular Weight:86.09 g/mol
- ADR:8,II
- :M-Clarity™ quality level = MQ200
- :Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
- UN:2531
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Solubility:98 g/l (20 °C)
- Melting Point:15 °C
- Molar Mass:86.09 g/mol
- Boiling Point:161 °C (1013 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:65 °C
- Refractive Index:1.431 (20 °C, 589 nm)
- Explosion Limit:1.6 - 8.7 %(V)
- Density:1.015 g/cm3 (20 °C)
- pH:2.0 - 2.2 (100 g/l, H2O, 20 °C)
- Ignition Point:370 °C
- Synonyms:2-Methylpropenoic acid
- Hill Formula:C4H6O2
Specifications
- Pack type:Glass Bottle
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



