PakSense™ XpressPDF™ Temperature Dataloggers, Thermco®
89412-414
: Avantor
- Interface:USB
- Description:55-Day Datalogger, with Alerts Above and Below 15 to 25°C
- Cat. no.:89412-414
- Supplier No.:XPDF551525-1
Designed to allow simplistic operation, PakSense™ XpressPDF™ Labels eliminate the need for reading devices and proprietary download software
These temperature loggers are optimal for applications that require fully independent operation, such as global shipments. Labels feature an integrated USB connection point which can be plugged directly into the USB port of a computer for data generation. Three options are available, including a highly accurate model for use in the life sciences industry, a general-use model, and a basic version.
: UL listed. CE marked.
: Accessories not included.