Specifications
- Description:Tube and Cap Strips, 100 µl Capacity
- Cat. no.:89428-994
- Supplier No.:T319-4N
Specifications
About this item
These RotoCycler™ strips and discs are specially made to be used with the Qiagen Rotor-Gene™ Q Real-Time Rotary Analyzer.
Tube strips are packaged separately from cap strips. The frosted extensions on caps not only make them more efficient and secure during handling, but also offer a convenient area for labelling. For individual use, tube and cap strips can easily be separated and used as individual units. Each package contains one bag of 250 tube strips and one bag of 250 cap strips. Case content is sufficient for 1000 reactions. 4 packages per case.
Two discs models are available: a 72-well format with 100 µL tubes and a 100-well format for reactions up to 25 µL. The discs are a one-piece "plate" equivalent, having vertically oriented wells compatible with automated reaction setup using a robotic liquid handling system.
The workstation especially made to facilitate the insertion of caps on the tubes. The rack can hold up to 18 x 0.1 ml tube strips for a total of 72 tubes. Other cavities can hold larger reaction tube.