Order Entry
Mexico
Orders LinkContactUsLinkComponent
Eppendorf LoBind® twin.tec® PCR Plates

Eppendorf LoBind® twin.tec® PCR Plates

25 plates (5 bags × 5 plates), 96, Fully skirted, 150 µl, Clear, Colourless
Catalog # 10049-106
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   EPPENDORF BIO TOOLS
Supplier #:  0030129512
Eppendorf LoBind® twin.tec® PCR Plates
Catalog # 10049-106
Product available on GSA Advantage®
   Low Manufacturing Impact
Eppendorf LoBind® twin.tec® PCR Plates
Eppendorf LoBind® twin.tec® PCR Plates

25 plates (5 bags × 5 plates), 96, Fully skirted, 150 µl, Clear, Colourless

Price
$4,800.00
Log in for availability
Paquete de 25
Product Details & Documents

Eppendorf twin.tec® PCR plates LoBind, available in 96 and 384-well formats, maximize nucleic acid recovery and are certified as 'PCR clean' for consistently reliable results.

  • LoBind material guarantees maximum sample recovery for improved assay results
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Available in 96- and 384-well formats, twin.tec® PCR plates LoBind® improve recovery of nucleic acids by reducing their adsorption to the wall of the wells. A combination of special manufacturing technologies and selected polypropylene batches ensures that nearly 100 % of DNA and RNA molecules are recovered without the use of surface coatings, thereby eliminating the risk of sample contamination.

Offering maximum confidence in results, all Eppendorf twin.tec PCR plates LoBind are'‘PCR clean'. This means the plates are batch-tested and certified by an independent laboratory to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors.

Autoclavable: 121 °C, 20 min.
Centrifugation stability: 6000×g
Purity grade: PCR clean (nonsterile)

Specifications
  • Frame Color:
    Colourless
  • No. of wells:
    96
  • Skirt:
    Fully skirted
  • Well Color:
    Clear
  • Well Volume:
    150 µl
  • Description:
    25 plates (5 bags × 5 plates)
  • Cat. No.:
    10049-106
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact