Zone Samplers, Bürkle
About this item
For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fine powder to coarse grain.
- Easy to clean, removable tip
- Colour coding (Multi and Uno samplers)
- Ultra-pure materials: Stainless steel V4A (1.4404)/AISI 316 L, PTFE or aluminium
- 3 lengths from 550 to 1500 mm, Jumbo up to 2500 mm
- Samplers have smallest possible gap between inner and outer tubes
Multi sampler has three or five separate chambers for taking samples at multiple depths in one action. Uno sampler has one chamber for taking a sample at one specified depth. All layer sampler has no chambers, three or five inlets and this allows one sample to be taken at multiple depths. Jumbo sampler has no chambers and three, five or seven inlets; with a 50 mm tube diameter this is ideal for larger grain sizes and quantities.
11 Options Available Below
Product Details & Documents
For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fine powder to coarse grain.
- Easy to clean, removable tip
- Colour coding (Multi and Uno samplers)
- Ultra-pure materials: Stainless steel V4A (1.4404)/AISI 316 L, PTFE or aluminium
- 3 lengths from 550 to 1500 mm, Jumbo up to 2500 mm
- Samplers have smallest possible gap between inner and outer tubes
Multi sampler has three or five separate chambers for taking samples at multiple depths in one action. Uno sampler has one chamber for taking a sample at one specified depth. All layer sampler has no chambers, three or five inlets and this allows one sample to be taken at multiple depths. Jumbo sampler has no chambers and three, five or seven inlets; with a 50 mm tube diameter this is ideal for larger grain sizes and quantities.
Product Family Options
All Layer Samplers
Product Information
Multi Samplers
Product Information
Uno Samplers
Product Information
Jumbo Samplers
Product Information
- LengthChamber VolumeInsertion DepthNo. of OpeningsAvailabilityPrice
- 10217-108All layer-open iiner tube, aluminiumSupplier Number: 5319-1085
- 10217-114Multi-closed chambers, Aluminium/PTFEMulti-closed chambers, Aluminium/PTFE95 cm390 ml850 mm3Log in for availabilityEach (1UN)$41,269.91Supplier Number: 5319-2085
- 10217-116Multi-closed chambers, Aluminium/PTFEMulti-closed chambers, Aluminium/PTFE165 cm650 ml1500 mm5Log in for availabilityEach (1UN)$48,545.09Supplier Number: 5319-2150
- 10217-118Multi-closed chambers, Aluminium/PTFEMulti-closed chambers, Aluminium/PTFE265 cm910 ml2500 mm7Log in for availabilityEach (1UN)$65,453.67Supplier Number: 5319-2250