SP Bel-Art Cleanware™ Scrubbing Sponge, Bel-Art Products, a part of SP
Catalog # 10147-770
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Cleanware™ Absorbent Sponge for Glassware
- Cat. no.:10147-770
- L×W×H:10.8×6.4×2.5 cm (4¼×2½×1")
- Supplier No.:F17078-0001
Specifications
About this item
Remove stubborn residue on glassware with this odor-resistant, highly absorbent sponge.
- Wipe dry performance equal to cellulose; maintains the desirable physical properties of polyurethane
- Foam contains anti-microbial technology to inhibit odor causing bacteria within the sponge itself
- Stays soft and flexible, wet or dry; will not crack or shrink, or become brittle like cellulose
- Scouring side resists snagging; does not leave fibers behind and retains aggressive scouring ability even with frequent use