Order Entry
Mexico
ContactUsLinkComponent
3B Scientific® BONElike™ Vertebrae Sets
3B Scientific® BONElike™ Vertebrae Sets
Catalog # 470118-990
undefined
3B Scientific® BONElike™ Vertebrae Sets
Catalog # 470118-990
Supplier Number:  1000156
Specifications
  • Description:
    Vertebrae Set
  • Cat. no.:
    470118-990
  • Supplier No.:
    A793
Specifications
About this item
This set is a worldwide unique original anatomical replica of a human vertebrae with the most precise illustration of even the finest anatomic structures of the vertebrae, exhibiting excellent quality.
  • Includes 3B smart anatomy
These vertebrae are excellent to use as a substitute for real bones for medical teaching purposes and patient consultation. The vertebrae set includes the 7 cervical, 12 thoracic and 5 lumbar vertebrae. Each vertebra is labeled for identification purposes (C1-7, T1-12 and L1-5).
The anatomical replica is supplied in a transport and storage case with individual compartments for all 24 vertebrae. Sets of individual regions are supplied in a contoured base.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.