Order Entry
Mexico
ContactUsLinkComponent
KIMBLE® Bleed Type Inlet Adapter with Hose Connection and Standard Taper Joint, DWK Life Sciences
KIMBLE® Bleed Type Inlet Adapter with Hose Connection and Standard Taper Joint, DWK Life Sciences
Catalog # KT179000-2440
undefined
KIMBLE® Bleed Type Inlet Adapter with Hose Connection and Standard Taper Joint, DWK Life Sciences
Catalog # KT179000-2440
Supplier Number:  179000-2440

Specifications

  • Cap Type:
    [ST] 24/40 with Hose Connection
  • Description:
    Adapter Gas Inlet, Plain Bottom, Tube Length 200 mm
  • Cat. no.:
    KT179000-2440
  • Overall Height:
    290 mm
  • Fits Tubing I.D.:
    ⅜"
  • Supplier No.:
    179000-2440

Specifications

About this item

Used to introduce reactants below the liquid surface.

  • Bleed-type design
  • [ST] joint
  • With hose connection

Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.