Order Entry
Mexico
ContactUsLinkComponent
Inoculation Loops, Calibrated, Disposable
Inoculation Loops, Calibrated, Disposable
Catalog # 30002-166
Inoculation Loops, Calibrated, Disposable
Catalog # 30002-166
Supplier Number:  COP-S10
Specifications
  • Description:
    Flexible (Soft) Plastic Loop
  • Size:
    10 μl
  • Cat. no.:
    30002-166
  • Packaging:
    200/box
  • Handle Color:
    Light Blue
  • Supplier No.:
    COP-S10
Specifications
About this item
Ultra smooth loop surfaces allows easy streaking
No rough plastic edges, flashing or burrs on the loop head means smooth, problem-free planting and streaking of cultures. Loops do not cut or gouge the agar surface during streaking. Free of lubricants, oils and electrostatic charges enabling consistent wetting and complete liquid transfer. Hexagonal loop shaft improves grip, assists orientation and makes diluting and streaking easy
Certifications: Loop accuracy certified using Evans Blue Dye Method. Certification of Calibration in every box.
Ordering information: Packaged as 25 loops per bag and 1,000 loops per box. Zip-lock closure reseals the pack between use to avoid contamination.