Order Entry
Mexico
ContactUsLinkComponent
PTFE-Lined Closed Caps, Thermo Scientific
PTFE-Lined Closed Caps, Thermo Scientific
Catalog # 46600-681
undefined
PTFE-Lined Closed Caps, Thermo Scientific
Catalog # 46600-681
Supplier Number:  B267-2414UBB
Specifications
  • Color:
    White
  • Description:
    Closed Top Cap with Bonded PTFE Liner
  • Material:
    PP
  • Cat. no.:
    46600-681
  • Supplier No.:
    B267-2414UBB
Specifications
About this item
PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials
PTFE-faced silicone liner provides added cushion and an extra secure seal when used with bottles and VOA vials.
The same high-quality PTFE-lined caps used on Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it will not fall out or become separated from the cap.