Solar-Powered Car
Solar-Powered Car
Catalog # 470117-792
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: SLRCR1
Jump to:
Product Details & Documents
This economical demonstration solar car can be powered by a solar cell or a rechargeable battery (AA size, not included).
- Economical
- Rechargable
- Includes two binding posts
Battery can be recharged from the solar cell, demonstrating an electricity storage system for solar power. The solar cell can be tilted to maximize solar input, features an on/off and select function switch, and includes two binding posts to measure output. All working parts are visible through the clear molded housing. Includes activity guide.
Documents
Specifications
- Description:Solar-Powered Car