Delta™ Cell Spreader
Delta™ cell spreader, Purple, 172 mm, Sterile
Supplier: HEATHROW SCIENTIFIC
: HS8151
Jump to:
Product Details & Documents
Flexible rippled handle and domed spreading bar helps reduce agar tears.
- Made from ABS
- Sterile
Documents
Certificates
Search by Lot NumberSpecifications
- Sterility:Sterile
- Color:Purple
- Description:Delta™ cell spreader
- Overall Length:172 mm