Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Potassium Standards
Potassium AAS Standards, RICCA Chemical Company
Potassium AAS Standards, RICCA Chemical Company
RCAK1KW-500
:
RICCA CHEMICAL CO
Small Business Enterprise
:
Potassium AAS Standards, RICCA Chemical Company
RCAK1KW-500
:
RICCA CHEMICAL CO
:
RAK1KW00-500N
Small Business Enterprise
:
Concentration:
1000 µg/ml
Matrix:
H₂O
Pack type:
Plastic bottle
Size:
500 ml
Cat. no.:
RCAK1KW-500
Element:
Potassium
Supplier No.:
RAK1KW00-500N