Datalogging Thermometer, 12-Channel, Extech
10068-326
: Avantor
- Description:12-Channel Datalogging Thermometer
- Cat. no.:10068-326
- Range:T: –100…+1150 °C (–148...2102 °F) (J)
T: –100…+1300 °C (–148…+2372 °F) (K)
T: –100…+400 °C (–148…+752 °F) (T)
T: –100…+900 °C (–148…+1652 °F) (E)
T: 0...1700 °C (32...3092 °F) (R)
T: 0...1500 °C (32...2732 °F) (S) - Supplier No.:TM500
Extech’s 12-Channel datalogging thermometer allows for a wide-range of applications with 6 Thermocouple types (J, K, E, T, R,S)
Offset adjustment used for zero function or to make relative measurements. Unit stores 99 readings manually and 20M readings via 2GB SD card.
It displays CH1 to CH8 or CH9 to CH12 on screen, and the data can be recorded from all 12 channels simultaneously onto an SD card in Excel format for further analysis.
: CE