Order Entry
Mexico
Orders LinkContactUsLinkComponent
Phenolphthalein ACS

Phenolphthalein ACS

Supplier:   VWR
Phenolphthalein ACS
Supplier:  VWR
Certificates
Phenolphthalein ACS

2 Options Available Below

Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS Number:77-09-8
  • Formula:C₂₀H₁₄O₄
  • MDL Number:MFCD00005913
  • Molecular Weight:318.33 g/mol
  • Merck Index:14,07243
  • Melting Pt:258 - 262 °C
  • Melting Pt:258…262 °C
  • Density:1.3 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-004-7
  • REACH:01-2119498295-24

Documents

Specifications for Product Family

Specification Test Results

  • Clarity of Alcohol Solution:
    Pass
  • Visual Transition Interval (pH 8-10):
    Pass
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Poly bottle
    50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $2,381.37
    $2,381.37
    Each (50g)
       $2,381.37

    Supplier Number: 0376-50G
  • Poly bottle
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $2,430.06
    $2,430.06
    Each (100g)
       $2,430.06

    Supplier Number: 0376-100G
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.